Learn More
PHYH Monoclonal Antibody (1F2-5B9), Invitrogen™
Mouse Monoclonal Antibody
Supplier:  Thermo Scientific MA519595
Catalog No. PIMA519595
Add to Cart
Description
Description
Peptide Sequence: MEQLRAAARL QIVLGHLGRP SAGAVVAHPT SGTISSASFH PQQFQYTLDN NVLTLEQRKF YEENGFLVIK NLVPDADIQR FRNEFEKICR KEVKPLGLTV MRDVTISKSE YAPSEKMITK VQDFQEDKEL FRYCTLPEIL KYVECFTGPN IMAMHTMLIN KPPDSGKKTS RHPLHQDLHY FPFRPSDLIV CAWTAMEHIS RNNGCLVVLP GTHKGSLKPH DYPKWEGGVN KMFHGIQDYE ENKARVHLVM EKGDTVFFHP LLIHGSGQNK TQGFRKAISC HFASADCHYI DVKGTSQENI EKEVVGIAHK FFGAENSVNL KDIWMFRARL VKGERTNL
PHYH Monoclonal antibody specifically detects PHYH in Human samples. It is validated for ELISA, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PHYH
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
O14832
Phyh
PHYH (AAH29512, 1 a.a. ∽ 338 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG1 κ
ELISA, Western Blot
1F2-5B9
Unconjugated
Phyh
1600020H07Rik; 2-HPCL; AI256161; AI265699; HPCL; HPCL2; induced by chronic shear stress; LN1; Lnap1; LOC100056651; Lupus nephritis-associated peptide 1; OTTHUMP00000019131; OTTHUMP00000019132; OTTHUMP00000179083; OTTHUMP00000216226; Pahx; PHYH; PHYH protein; PHYH1; PHYH2; phytanic acid oxidase; phytanoil-CoA alpha hydroxylase; phytanoyl-CoA 2 oxoglutarate dioxygenase; phytanoyl-CoA 2-hydroxylase; phytanoyl-CoA alpha-hydroxylase; phytanoyl-CoA dioxygenase peroxisomal; phytanoyl-CoA dioxygenase, peroxisomal; phytanoyl-CoA hydroxylase; phytanoyl-CoA hydroxylase (Refsum disease); RD; zgc:110203
Mouse
Affinity chromatography
RUO
5264
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions