Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHYH Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526017

Catalog No. PIPA526017


Add to cart

Description

Description

PHYH2 catalyzes a carbon-carbon cleavage reaction; cleaves a 2-hydroxy-3-methylacyl-CoA into formyl-CoA and a 2-methyl-branched fatty aldehyde.
Specifications

Specifications

PHYH
Polyclonal
PBS with 0.09% sodium azide
O14832
2-HPCL, HPCL, HPCL2, PHYH2, 1600020H07Rik, 2-hydroxyphytanol-CoA lyase, 2-hydroxyphytanoyl-CoA lyase, phytanoyl-CoA 2-hydroxylase 2, phytanoyl-CoA hydroxylase 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5264
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
PHYH
Liquid
PHYH
KLH conjugated synthetic peptide between 59-87 amino acids from the N-terminal region of human PHYH
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit