Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3R1 Rabbit anti-Human, Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA129194

Catalog No. PIPA129194

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-29195 detects Phospholipase C1 from bovine samples. PA1-29195 has been successfully used in immunoprecipitation, Western blot applications. The PA1-29195 immunogen is: Synthetic peptide corresponding to the C-terminus of Bovine PLCgamma1 protein

Phosphatidylinositol 3-kinase (PIK3) is a signaling protein that phosphorylates the inositol ring of phosphatidylinositol at the 3-prime position. Signaling is initiated when upstream tyrosine kinase receptors or G protein coupled receptors are bound by their ligands. It binds to activated (phosphorylated) protein-Tyr kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane. Phosphatidylinositol 3-kinase plays an important role in the metabolic actions of insulin, and a mutation in this gene has been associated with insulin resistance. During insulin stimulation, it also binds to IRS-1. Mutations in the gene can result in Agammaglobulinemia 7, SHORT syndrome, or Immunodeficiency 36.
Specifications

Specifications

PIK3R1
Polyclonal
Unconjugated
PIK3R1
Liquid
Rabbit
IgG
500 μL
4° C, do not freeze
Primary
25513, 5295
Western Blot
1 mg/mL
PBS with 0.2% BSA and 0.1% sodium azide; pH 7.4
P27986, Q63787
PIK3R1
Synthetic peptide from the C terminal SH2 domain of p85 alpha subunit of phosphatidylinositol (PI) 3-kinase.
Protein A
RUO
Antibody
Polyclonal
Human, Non-human Primate, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit