Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PI 4 Kinase type 2 beta, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132263

Catalog No. NBP132263


Add to cart

Description

Description

PI 4 Kinase type 2 beta Polyclonal antibody specifically detects PI 4 Kinase type 2 beta in Human samples. It is validated for Western Blot.
Specifications

Specifications

PI 4 Kinase type 2 beta
0.72mg/mL
Western Blot 1:500-1:3000
Affinity Purified
PI4K2B
Recombinant protein encompassing a sequence within the center region of human PI4K2B. The exact sequence is proprietary.
55 kDa
0.1 ml
Protein Kinase
Primary
55300
Human
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
EC 2.7.1.67, FLJ11105, phosphatidylinositol 4-kinase type 2 beta, phosphatidylinositol 4-kinase type 2-beta, Phosphatidylinositol 4-kinase type II-beta, phosphatidylinositol 4-kinase type-II beta, PI4KIIB, PI4KII-BETA, PIK42B
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only