Learn More
PI3 Kinase-C2-alpha Mouse anti-Mouse, Rat, Human, Clone: 9H4.2, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051560
Catalog No. 05-156-0MI
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-PI3 Kinase-C2-alpha Antibody, clone 9H4.2 detects level of PI3 Kinase-C2-alpha and has been published and validated for use in western blotting and immunocytochemistry.
Specifications
Specifications
PI3 Kinase-C2-alpha
Monoclonal
Unconjugated
NP_002636.2
Mouse
Ion Exchange
Primary
2-8°C for one year
IgM κ
Immunocytochemistry, Western Blot
9H4.2
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
CPK, PI3K-C2-alpha, PI3-K-C2(ALPHA), PI3-K-C2A, PIK3C2A
KLH-conjugated linear peptide corresponding to human PI3K-C2-alpha.
100 μg
Human, Mouse, Rat
Purified