Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3C2A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528071

Catalog No. PIPA528071


Add to cart

Description

Description

PA5-28071 targets PI3 kinase C2 alpha in IF, IHC-P, and WB applications and shows reactivity with Human samples. The PA5-28071 immunogen is synthetic peptide corresponding to a region within amino acids 1624 and 1686 of Human PI3 kinase C2 alpha. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene belongs to the phosphoinositide 3-kinase (PI3K) family. PI3-kinases play roles in signaling pathways involved in cell proliferation, oncogenic transformation, cell survival, cell migration, and intracellular protein trafficking. This protein contains a lipid kinase catalytic domain as well as a C-terminal C2 domain, a characteristic of class II PI3-kinases. C2 domains act as calcium-dependent phospholipid binding motifs that mediate translocation of proteins to membranes, and may also mediate protein-protein interactions. The PI3-kinase activity of this protein is not sensitive to omolar levels of the inhibitor wortmanin. This protein was shown to be able to be activated by insulin and may be involved in integrin-dependent signaling.
Specifications

Specifications

PIK3C2A
Polyclonal
Unconjugated
PIK3C2A
Liquid
PIK3C2A
Synthetic peptide corresponding to a region within amino acids 1624 and 1686 of Human PI3 kinase C2 alpha
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.8 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O00443
CPK, PI3-K-C2(ALPHA), PI3-K-C2A, C2-containing phosphatidylinositol kinase, PI3K-C2-alpha, PI3K-C2alpha, phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit alpha, phosphoinositide 3-kinase-C2-alpha, ptdIns-3-kinase C2 subunit alpha
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5286
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit