Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI3-Kinase Mouse, Unlabeled, Clone: 4, BD

Mouse Monoclonal Antibody

$211.00 - $448.00

Specifications

Antigen PI3-Kinase
Isotype IgG2a
Immunogen Human PI3-Kinase α subunit aa. 562 to 724
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610046
View Documents
BD Biosciences
610046
150μg
Each for $448.00
Add to cart
 
BDB610045
View Documents
BD Biosciences
610045
50μg
Each for $211.00
Add to cart
 
Description

Description

PI3-kinase phosphorylates the D-3 position of the inositol ring of phosphatidylinositol (PtdIns), PtdIns(4)P and PtdIns(4,5)P2 to produce the respective PI3-phosphorylated derivatives. PI3-kinase exists as a heterodimer of 85 kDa (p85) and 110 kDa (p110) subunits. The p85 subunit contains two SH2 domains and an SH3 domain. It associates with and serves as a substrate for activated growth factor receptor tyrosine kinases. p85 may serve as regulator of the catalytic subunit, p110, by acting as the link between PI3-kinase and the ligand-activated receptor. Two distinct forms of the p85 subunit have been described: 1) p85 α; , which binds tightly to the catalytic subunit, and 2) p85ß, a protein whose function is presently unknown. Both isoforms bind to activated receptors and serve as tyrosine kinase substrates.
Specifications

Specifications

PI3-Kinase
Human PI3-Kinase α subunit aa. 562 to 724
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG2a
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
4
250μg/mL
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit