Learn More
PI3 Kinase Mouse anti-Human, Clone: AW103, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05703
Catalog No. 05-703-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This Anti-PI3 Kinase Antibody, p110δ, clone AW103 is validated for use in western blotting for the detection of PI3 Kinase.
Specifications
Specifications
PI3 Kinase
AW103
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
κ
200μg
Primary
Human
Western Blot
Unconjugated
NM_005026
PIK3CD, p110d, p110delta, PI3K, p110D
GST fusion protein corresponding to residues 270-334 of human PI3 Kinase p110 delta
Protein G purfied
−20°C for 24 months.
Monoclonal