Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3CB Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522152

Catalog No. PIPA522152


Add to cart

Description

Description

PA5-22152 targets PI3 Kinase p110 beta in ICC, IF, and WB applications and shows reactivity with Human and mouse samples. The PA5-22152 immunogen is a recombit fragment corresponding to a region within amino acids 509 and 820 of PI3 kinase p110 beta (Uniprot IDNo.P42338). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

PIK3CB
Polyclonal
Unconjugated
PIK3CB
Liquid
PIK3CB
Recombit fragment corresponding to a region within amino acids 509 and 820 of PI3 kinase p110 beta (Uniprot IDNo.P42338)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P42338, Q8BTI9, Q9Z1L0
P110BETA, PI3K, PI3KBETA, PIK3C1, PI3-kinase p110 subunit beta, PI3-kinase subunit beta, PI3K-beta, PtdIns-3-kinase p110, phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit beta, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta isoform, ptdIns-3-kinase subunit beta, ptdIns-3-kinase subunit p110-beta
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5291, 74769, 85243
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit