Learn More
PI3-kinase type 3 Mouse anti-Human, Mouse, Rat, Clone: 17D1.1, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051563
Catalog No. 05-156-3MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-PI3-kinase type 3 Antibody, clone 17D1.1 detects level of PI3-kinase type 3 and has been published and validated for use in western blotting and immunocytochemistry.
Specifications
Specifications
PI3-kinase type 3
17D1.1
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Purified
Murine
IgM, κ, κ
100μg
Primary
Human, Murine, Rat
Immunocytochemistry, Western Blot
Unconjugated
NP_002638
PIK3C3, VPS34
KLH-conjugated linear peptide corresponding to human PI3-kinase type 3.
Affinity purified
2-8°C for one year
Monoclonal