Learn More
PI4-Kinase Β Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06578
Catalog No. 06-578-MI
Add to cart
Description
Description
Detect PI4-Kinase β using this Anti-PI4-Kinase β Antibody validated for use in immunocytochemistry, immunoprecipitation and western blotting.
Specifications
Specifications
PI4-Kinase β
Unconjugated
Purified
Rabbit
IgG
200μg
Primary
NM_006215
Immunocytochemistry, Immunoprecipitation, Western Blot
0.1M Tris-glycine, pH 7.4, 0.15M NaCl with 0.05% Sodium Azide
PIK4CB; PI4KIIIbeta; PI4K-beta; PI4Kbeta; NPIK; PI4K-BETA; PI4KB; PI4K92
GST fusion protein corresponding to residues 410-537 of human PI 4-Kinase beta
Protein A chromatography
−20°C for 24 months
Polyclonal
Human, Murine, Rat