Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PIASy, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP177161

Catalog No. NBP177161

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

PIASy
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
NP_056981
E3 SUMO-protein ligase PIAS4, FLJ12419, MGC35296, Piasg, PIAS-gamma, PIASy, Protein inhibitor of activated STAT protein 4, Protein inhibitor of activated STAT protein gamma, protein inhibitor of activated STAT protein PIASy, protein inhibitor of activated STAT, 4, zinc finger, MIZ-type containing 6, ZMIZ6
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
51588
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Affinity Purified
PIAS4
Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human PIAS4. The immunogen is located within amino acids 50 - 100 of PIAS4.
56 kDa
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only