Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-PIBF1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP219823
Description
PIBF1 Polyclonal specifically detects PIBF1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PIBF1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
C13orf24, chromosome 13 open reading frame 24, KIAA1008, PIBF, progesterone immunomodulatory binding factor 1, progesterone-induced blocking factor 1, progesterone-induced-blocking factor 1, RP11-505F3.1 | |
Rabbit | |
90 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (92%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
Q8WXW3 | |
PIBF1 | |
Recombinant protein encompassing a sequence within the C-terminus region of human PIBF1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
10464 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only