Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pICIn Mouse, Unlabeled, Clone: 32, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611274

Catalog No. BDB611274

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

pICln (I = current, Cl = chloride, n = nucleotide-sensitive) was initially described as an integral component of a nucleotide-sensitive chloride channel. However, it lacks homology to known channel proteins and exhibits cytoplasmic and nuclear localization. Thus, pICln is thought to be a cytosolic regulator of a swelling-induced chloride channel, rather than a component of a channel itself. In support of this hypothesis, pICln interacts with several cytosolic proteins. It binds to several Sm proteins, which are common spliceosomal core proteins whose assembly
is necessary for the biogenesis of small nuclear ribonucleoproteins (snRNPs). Sm protein association with pICln inhibits their assembly. pICln also interacts with the 72kDa human homolog of Skb1, a yeast protein that binds a homolog of p21[Cdc42/Rac]-activated protein kinases. In addition, pICln interacts with, and is phosphorylated by, a constitutively active cytosolic protein kinase. Therefore, pICln is thought to be a component of signaling pathways that function in transcriptional or translational regulation, cytoskeletal reorganization, or a number of other biological processes.

Host Species: Mouse
Clone: 32
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human pICln aa. 92-201
Formula Weight [Chemical]: 37/42kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

pICIn
32
Unconjugated
Murine
IgG1
RUO
Primary
Canine, Human, Murine, Rat
Immunofluorescence, Western Blot
250μg/mL
Affinity Purified
Human pICln aa. 92-201
50μg
Cell Biology
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit