Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PICK1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA513841
Description
This antibody is predicted to react with bovine, non-human primate and rat based on sequence homology.
PSD-95/DLG/ZO-1 (PDZ) domain-containing proteins play a central role in synaptic membrane protein localization. The protein interacting with protein kinase C (PICK1) is a synaptic PDZ domain protein that also contains a coiled-coil and acidic domain. Studies indicate that PICK1 functions as a targeting and transport protein and has been shown to interact with protein kinase C alpha (PKC alpha), AMPA-type glutamate receptors, and several other membrane receptors via its PDZ domain. The interaction of PICK1 with PKC alpha is highly dependent on the activation of the kinase. It appears that PICK1 directs the activated form of PKC alpha to membrane anchored GluR2. Once phosphorylated by PKC alpha, GluR2 is released from the synaptic anchor proteins. Once released, GluR2 is transported from the synaptic membrane in a PICK1-dependent manner.Specifications
PICK1 | |
Polyclonal | |
PICK1 | |
PRKCA-binding protein; Protein kinase C-alpha binding protein; Protein interacting with C kinase 1; PICK1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
18693, 9463 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q9NRD5, Q62083 | |
PICK1 | |
KLH conjugated synthetic peptide between 286-316 amino acids from the C-terminal region of human PICK1 (PRKCABP) | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title