Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIF1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP197931
Description
PIF1 Polyclonal specifically detects PIF1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC/IF (Negative).Specifications
PIF1 | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
C15orf20, PIF, PIF1 5'-to-3' DNA helicase homolog (S. cerevisiae), PIF1 5-to-3 Prime DNA helicase homolog, PIF1 DNA helicase, PIF1/RRM3 DNA helicase-like protein, RRM3 DNA helicase-like protein | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Mouse | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.16 mg/mL | |
Immunohistochemistry 1:300, Immunohistochemistry-Paraffin 1:300, Western Blot (Negative), ICC/IF (Negative) | |
Q9H611 | |
PIF1 | |
A synthetic peptide made to an internal portion of the human PIF1 protein (between residues 400-500) [UniProt Q9H611] | |
0.1 mL | |
Cell Cycle and Replication | |
80119 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction