Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIN Mouse, Unlabeled, Clone: 4, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610727

 View more versions of this product

Catalog No. BDB610727


Add to cart

Description

Description

PIN (protein inhibitor of nNOS) is a small protein of 89 amino acids initially described as a light chain subunit of dynein and as an inhibitor of the neuronal nitric oxide synthase isoform (nNOS). In vitro, PIN has been reported to bind to a unique nNOS domain encompassing amino acids 163-245. PIN inhibits nNOS activity and blocks the formation of the active nNOS dimer. Although ubiquitously expressed, PIN levels are reportedly highest in brain and testis. Immunolocalization studies in Drosophila melanogaster detected a cytoplasmic distribution of PIN. Partial-loss-of-function of PIN resulted in morphologic and developmental changes in the bristles, wings, and female sterility of D. melanogaster. Deletion of PIN resulted in lethality with the characteristic morphology of apoptotic cells.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

PIN
4
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Immunofluorescence, Western Blot
250μg/mL
Protein Inhibitor of nNOS
Affinity Purified
Rat PIN aa. 1-89
150μg
Cell Biology
Primary
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.