Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PINCH Mouse anti-Human, Unlabeled, Clone: 49, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612710

 View more versions of this product

Catalog No. BDB612710


Add to cart

Description

Description

Integrins are transmembrane receptors that mediate cell-cell or cell-matrix adhesion. All integrins are heterodimers composed of α and β subunits, which interact with extracellular matrix and cytoskeletal proteins. Signal transduction through intergin receptors may be regulated by integrin-linked kinase (ILK). ILK is a widely expressed Ser/Thr protein kinase that contains four ankyrin-like repeats in the N-terminal region, a phoshoinositide lipid-binding motif at amino acids 180-212, and an integrin binding site at amino acids 293-451. The ankyrin repeats interact with a LIM domain-only protein called PINCH that also binds the adaptor protein, Nck-2. This interaction implicates ILK in growth factor receptor pathways. The PINCH adaptor protein also contains a leucine-rich nuclear export signal and a nuclear localization signal. These two motifs implicate PINCH in nuclear functions, and in Schwann cells PINCH has been found in the nucleus, as well as, cytoplasm and perinuclear regions. Thus, PINCH is an adaptor protein that functions in both integrin signaling and nuclear functions.

Immunofluorescence, Western Blotting

Specifications

Specifications

PINCH
49
Unconjugated
Affinity Purified
Human PINCH aa. 120-220
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.