Learn More
anti-PINCH1/LIMS1, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP131220
Catalog No. NBP131220
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
PINCH1/LIMS1
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
LIMS1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human LIMS1. The exact sequence is proprietary.
37 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Bovine (89%). Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
LIM and senescent cell antigen-like domains 1, LIM and senescent cell antigen-like-containing domain protein 1, PINCH-1, PINCHPINCH1Particularly interesting new Cys-His protein 1, Renal carcinoma antigen NY-REN-48
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
3987
Mouse, Rat