Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP5Kγ Mouse, Unlabeled, Clone: 12, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611148

Catalog No. BDB611148


Add to cart

Description

Description

Phosphoinositide turnover is a well established mechanism of intracellular signal transduction. Sequential phosphorylation of phosphatidylinositol (PtdIns) results in PtdIns(4)P (PIP) and PtdIns(4,5)P2 (PIP2). Phospholipase C (PLC) hydrolyzes PIP2 to inositol (1,4,5)P3 (IP3) which stimulates release of intracellular Ca2+. PIP2 is generated by phosphorylation of PtdIns 5-kinases (PI5-K). These enzymes are divided into two types (I and II) based on their size and sensitivity to certain compounds. Three mammalian PI5-Ks, PI5-Kα, β, and γ of type I have been identified and a type II PIP5kα. Although the PI4-Ks are abundantly distributed throughout the cell, activity is found primarily in association with membranous structures. Members of this family contain a lipid kinase unique domain and a C-terminal catalytic domain.

Host Species: Mouse
Clone: 12
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse PIP5Kγ aa. 479-580
Formula Weight [Chemical]: 87-90kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

PIP5Kγ
12
Unconjugated
Affinity Purified
Mouse PIP5Kγ aa. 479-580
50μg
Cell Biology
Primary
Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit