Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP5K1C Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528274

Catalog No. PIPA528274


Add to cart

Description

Description

PA5-28274 targets PIP5K1C in WB applications and shows reactivity with Human and mouse samples. The PA5-28274 immunogen is recombit fragment corresponding to a region within amino acids 152 and 375 of Human PIP5K1C. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a member of the type I phosphatidylinositol-4-phosphate 5-kinase family of enzymes. A similar protein in mice is found in synapses and focal adhesion plaques, and binds the FERM domain of talin through its C-terminus.
Specifications

Specifications

PIP5K1C
Polyclonal
Unconjugated
PIP5K1C
Liquid
PIP5K1C
Recombit fragment corresponding to a region within amino acids 152 and 375 of Human PIP5K1C
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.49 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O60331, O70161
LCCS3, PIP5K-GAMMA, PIP5Kgamma, PIPKIg_v4, Human homolog of mouse phosphatidylinositol-4-phosphate 5-kinase I-gamma, PIP5K1-gamma, diphosphoinositide kinase, phosphatidylinositol-4-phosphate 5-kinase type-1 gamma, ptdIns(4)P-5-kinase 1 gamma, type I PIP kinase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
18717, 23396
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit