Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PIP5K2 gamma, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192272

Catalog No. NBP192272


Add to cart

Description

Description

PIP5K2 gamma Polyclonal antibody specifically detects PIP5K2 gamma in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

PIP5K2 gamma
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
EC 2.7.1, EC 2.7.1.149, FLJ22055, phosphatidylinositol-4-phosphate 5-kinase, type II, gamma, Phosphatidylinositol-5-phosphate 4-kinase type II gamma, phosphatidylinositol-5-phosphate 4-kinase type-2 gamma, phosphatidylinositol-5-phosphate 4-kinase, type II, gamma, PI(5)P 4-kinase type II gamma, PIP4KII-gamma, PIP5K2C
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Affinity Purified
PIP4K2C
This antibody was developed against Recombinant Protein corresponding to amino acids:LEKLKRDVEFLVQLKIMDYSLLLGIHDIIRGSEPEEEAPVREDESEVDGDCSLTGPPALVGSYGTSPEGIGGYIHSHRPLGPGEFESFIDVYAIRSAEGAPQKEVYFMGLI
Immunogen affinity purified
RUO
Primary
79837
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only