Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phosphatidylinositol-5-phosphate 4-kinase, type II, alpha, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human PIP5K2A protein.

Manufacturer:  Abnova Corporation H00005305B01

Catalog No. 89-005-962


Add to cart

Description

Description

Phosphatidylinositol-5,4-bisphosphate, the precursor to second messengers of the phosphoinositide signal transduction pathways, is thought to be involved in the regulation of secretion, cell proliferation, differentiation, and motility. The protein encoded by this gene is one of a family of enzymes capable of catalyzing the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-5,4-bisphosphate. The amino acid sequence of this enzyme does not show homology to other kinases, but the recombinant protein does exhibit kinase activity. This gene is a member of the phosphatidylinositol-5-phosphate 4-kinase family. [provided by RefSeq

Sequence: MATPGNLGSSVLASKTKTKKKHFVAQKVKLFRASDPLLSVLMWGVNHSINELSHVQIPVMLMPDDFKAYSKIKVDNHLFNKENMPSHFKFKEYCPMVFRNLRERFGIDDQDFQNSLTRSAPLPNDSQARSGARFHTSYDKRYIIKTITSEDVAEMHNILKKYHQYIVECHGITLLPQFLGMYRLNVDGVEIYVIVTRNVFSHRLSVYRKYDLKGSTVAREASDKEKAKELPTLKDNDFINEGQKIYIDDNNKKVFLEKLKKDVEFLAQLKLMDYSLLVGIHDVERAEQEEVECEENDGEEEGESDGTHPVGTPPDSPGNTLNSSPPLAPGEFDPNIDVYGIKCHENSPRKEVYFMAIIDILTHYDAKKKAAHAAKTVKHGAGAEISTVNPEQYSKRFLDFIGHILT
Specifications

Specifications

phosphatidylinositol-5-phosphate 4-kinase, type II, alpha
Unconjugated
No additive
NM_005028.3
FLJ13267|PI5P4KA|PIP5K2A|PIP5KII-alpha|PIP5KIIA|PIPK
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5305
Western Blot
Mouse polyclonal antibody raised against a full-length human PIP5K2A protein.
PIP4K2A
Serum
PIP4K2A
PIP5K2A (NP_005019.2, 1 a.a. ~ 406 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit