Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP5K1C Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515284

Catalog No. PIPA515284

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PIP5K1G is a member of the type I phosphatidylinositol-4-phosphate 5-kinase family of enzymes. A similar protein in mice is found in synapses and focal adhesion plaques, and binds the FERM domain of talin through its C-terminus.
Specifications

Specifications

PIP5K1C
Polyclonal
PBS with 0.09% sodium azide
O60331
Phosphatidylinositol-4-phosphate 5-kinase, type I, gamma; PIP5K1C
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23396
Immunohistochemistry, Western Blot
Unconjugated
PIP5K1C
Liquid
PIP5K1C
KLH conjugated synthetic peptide between 637-668 amino acids from the C-terminal region of human PIP5KI gamma (PIP5K1G)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit