Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP5K1A, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528215

Catalog No. PIPA528215

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Participates in the biosynthesis of phosphatidylinositol-4,5-bisphosphate. Mediates RAC1-dependent reorganization of actin filaments. Contributes to the activation of PLD2.

PA5-28215 targets PIPK I alpha in WB applications and shows reactivity with Human samples.The PA5-28215 immunogen is recombinant fragment corresponding to a region within amino acids 249 and 562 of Human PIPK I alpha.
Specifications

Specifications

PIPK I alpha
Polyclonal
Unconjugated
PIP5K1A
Purified
PIP5K1A
Recombinant fragment corresponding to a region within amino acids 249 and 562 of Human PIPK I alpha
Antigen affinity chromatography
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
8394
Western Blot
0.46mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q99755
68kDa type I phosphatidylinositol-4-phosphate 5-kinase alpha, PIP5K1-alpha, PIP5KIalpha, phosphatidylinositol-4-phosphate 5-kinase type-1 alpha, ptdIns(4)P-5-kinase 1 alpha
Rabbit
IgG
100μL
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit