Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PITPnm (Phosphatidylinositol transfer protein; Nir2; MrdgBα) Mouse, Unlabeled, Clone: 8, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612027

 View more versions of this product

Catalog No. BDB612027


Add to cart

Description

Description

PITPs (phosphatidylinositol transfer protein) are abundant cytosolic or transmembrane proteins that are required for phospholipase C (PLC) signaling and vesicular trafficking. All of the mammalian PITPs are homologous to Drosophila retinal degeneration B (rdgB) proteins, which are PITPs that are involved in retinal and olfactory neurosensory signaling, and mutations in these proteins lead to retinal degeneration. There are at least 5 mammalian PITPs, three cytosoloic PITPs, PITPα, PITPβ, and MrdgBβ, and two transmembrane PITPs: PITPnm (Nir2/MrdgBα) and Nir3. PITPα and β participate in PLC signaling, mediated by the ability of PITP to deliver PI to the signaling complex containing PI4 Kinase, PIP-5-Kinase, and the activated receptor. PITPnm contains an N-terminal PITP domain adjacent to a Ca2+-binding domain (CBD), and a C-terminal PYK2 binding domain. PITPnm mRNA is widely expressed with the highest expression in brain, and PITPnm protein is found in the ER and Golgi. Thus, PITPnm may be important for phosphoinositide synthesis in the Golgi, and may interact with PYK2 during activation of PI-4 signaling pathways.

Host Species: Mouse
Clone: 8
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Mouse PITPnm

Western Blotting

Specifications

Specifications

PITPnm (Phosphatidylinositol transfer protein; Nir2; MrdgBα)
8
Unconjugated
Murine
IgG1
RUO
Primary
Murine, Rat
Immunofluorescence, Western Blot
250μg/mL
Affinity Purified
Mouse PITPnm
150μg
Cell Biology
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.