Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PITPNM3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131070

Catalog No. NBP131070


Add to cart

Description

Description

PITPNM3 Polyclonal antibody specifically detects PITPNM3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PITPNM3
1.0mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, KnockDown Validated
Q9BZ71
membrane-associated phosphatidylinositol transfer protein 3, MGC157740, MGC157741, NIR-1, NIR1CORD5, Phosphatidylinositol transfer protein, membrane-associated 3, PITPnm 3, PITPNM family member 3, Pyk2 N-terminal domain-interacting receptor 1, RDGBA3, retinal degeneration B alpha 3
Rabbit
IgG
Antigen Affinity-purified
RUO
Primary
83394
Human
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
PITPNM3
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PITPNM3. The exact sequence is proprietary.
107 kDa
0.1 ml
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human. Predicted cross-reactivity based on sequence identity: Mouse (100%).
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only