Learn More
PKD1L2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSP00007W
Catalog No. PIOSP00007W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
This gene encodes a member of the polycystin protein family. The encoded protein contains 11 transmembrane domains, a latrophilin/CL-1-like GPCR proteolytic site (GPS) domain, and a polycystin-1, lipoxygenase, alpha-toxin (PLAT) domain. This protein may function as a component of cation channel pores. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
PKD1L2
Polyclonal
whole serum with no preservative
Q7Z442
Polycystin-1L2, PC1-like 2 protein, PKD1L2, PC1L2
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
PKD1L2
Lyophilized
PKD1L2
A synthetic peptide from the 7th extracellular loop of human Polycystic kidney disease protein 1-like 2
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
114780