Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKBα/Akt Mouse, Unlabeled, Clone: 7, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610837

 View more versions of this product

Catalog No. BDB610837


Add to cart

Description

Description

Rac (Related to the A and C kinases) protein kinase shows 73% and 68% to Protein Kinase C and PKA, respectively, and is also known as Protein Kinase B (PKB). It was also identified as the product of the v-akt oncogene from T-cell lymphomas. PKB mRNA is expressed in a wide variety of cell lines and has been detected in porcine brain, heart, liver, kidney, muscle, and ovary. Rac phosphorylates a number of physiological substrates including MBP, glycogen synthetase, PKA RII subunit, and histone H1. PKB is activated in response to growth factors through the activation of PI3-kinase and Ras. Activation of PI3-Kinase generates phosphatidylinositol 3,4-bisphosphate which may induce the membrane translocation of PKB coincident with its phosphorylation and activation. Cellular stress leading to the activation of the p38 MAP kinase also induces PKB activation, indicating a multiplicity of signaling pathways that activate PKB. Upon activation, PKB associates with some members of the PKC family of kinases, such as PKCδ and PKCζ. Downstream of PKB, GSK3 is thus far the only identified substrate for PKB phosphorylated in vivo.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

PKBα/Akt
7
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
PKBα
Affinity Purified
Human PKBα/Akt aa. 71-184
150μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.