Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKCε Mouse, Unlabeled, Clone: 21, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610086

 View more versions of this product

Catalog No. BDB610086


Add to cart

Description

Description

The Protein Kinase C (PKC) family of homologous serine/threonine protein kinases is involved in a number of processes such as growth, differentiation, and cytokine secretion. At least eleven isozymes have been described. These proteins are products of multiple genes and alternative splicing. PKC consists of a single polypeptide chain containing four conserved regions (C) and five variable regions (V). The N-terminal half containing C1, C2, V1, and V2 constitutes the regulatory domain and interacts with PKC activators Ca2), phospholipid, diacylglycerol, or phorbol ester. However, the novel PKC (nPKC) subfamily members (δ, η, η, and θ isoforms) and the atypical PKC (aPKC)
subfamily members (ζ , ι , and λ isoforms) are Ca(2+) independent and lack the C2 domain. The PKC pathway represents a major signal transduction system that is activated following ligand-stimulation of transmembrane receptors by hormones, neurotransmitters and growth factors. Expression of the 90kDa PKCα is induced by interferon-α. Generally, PKCα is expressed at very low levels in normal murine tissues, except brain. Overexpression of PKCα leads to increased growth rates and higher cell densities in monolayer cultures. A high level of expression is seen in several hematopoietic cell lines and tumors. This suggests a possible role for PKCα in tumorigenesis.

Host Species: Mouse
Clone: 21
Isotype: IgG2a
Species Reactivity: Rat
Immunogen: Human PKCε aa. 1-175
Formula Weight [Chemical]: 90kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

PKCε
21
Unconjugated
Affinity Purified
Human PKCε aa. 1-175
150μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.