Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKCβ Mouse, Unlabeled, Clone: 36, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610127

 View more versions of this product

Catalog No. BDB610127


Add to cart

Description

Description

The Protein Kinase C (PKC) family of homologous serine/threonine protein kinases is involved in a number of processes such as growth, differentiation, and cytokine secretion. At least eleven isozymes have been described. These proteins are products of multiple genes and alternative splicing. PKC consists of a single polypeptide chain containing four conserved regions (C) and five variable regions (V). The N-terminal half containing C1, C2, V1, and V2 constitutes the regulatory domain and interacts with the PKC activators Ca2+;, phospholipid, diacylglycerol, or phorbol ester. However, the novel PKC (nPKC) subfamily members ( δ, ε, η, and θ isoforms) and the atypical PKC (aPKC) subfamily members (ζ, ι, and λ isoforms) are Ca2+; independent and lack the C2 domain. The aPKC members are unique in that their activity is
independent of diacylglycerols and phorbol esters. They also lack one repeat of the cysteine-rich sequences that are conserved in cPKC and nPKC. The C-terminal region of PKC contains the catalytic domain. The PKC pathway represents a major signal transduction system that is activated following ligand-stimulation of transmembrane receptors by hormones, neurotransmitters and growth factors. PKCβ is highly expressed in brain and hematopoietic cells. Autophosphorylation of PKCβ occurs at the N- and C-terminal regions, as well as within the hinge region. However, only the COOH- terminal autophosphorylation sites are essential for PKCβ's function and subcellular localization. PKCβ is critical for the proliferation of K562 cells, as well as being an important regulator of human melanogenesis.

Host Species: Mouse
Clone: 36
Isotype: IgG2a
Species Reactivity: Rat
Immunogen: Human PKCβ aa. 126-324
Formula Weight [Chemical]: 80kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

PKCβ
36
Unconjugated
Affinity Purified
Human PKCβ aa. 126-324
50μg
Cell Biology
Primary
Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.