Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC lambda/iota, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527811

Catalog No. PIPA527811

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a member of the protein kinase C (PKC) family of serine/threonine protein kinases. The PKC family comprises at least eight members, which are differentially expressed and are involved in a wide variety of cellular processes. This protein kinase is calcium-independent and phospholipid-dependent. It is not activated by phorbolesters or diacylglycerol. This kinase can be recruited to vesicle tubular clusters (VTCs) by direct interaction with the small GTPase RAB2, where this kinase phosphorylates glyceraldehyde-3-phosphate dehydrogenase (GAPD/GAPDH) and plays a role in microtubule dynamics in the early secretory pathway. This kinase is found to be necessary for BCL-ABL-mediated resistance to drug-induced apoptosis and therefore protects leukemia cells against drug-induced apoptosis. There is a single exon pseudogene mapped on chromosome X.

PA5-27811 targets PKC iota in IF, IHC (P), and WB applications and shows reactivity with Human samples.The PA5-27811 immunogen is recombinant fragment corresponding to a region within amino acids 1 and 233 of Human PKC iota.
Specifications

Specifications

PKC iota
Polyclonal
Unconjugated
PRKCI
Purified
PRKCI
Recombinant fragment corresponding to a region within amino acids 1 and 233 of Human PKC iota
Antigen affinity chromatography
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
5584
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.5mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P41743
DXS1179E, PKCI, nPKC-iota, PRKC-lambda/iota, aPKC-lambda/iota, atypical protein kinase C-lambda/iota, protein kinase C iota type
Rabbit
IgG
100μL
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit