Learn More
PKD1L1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSP00008W
Catalog No. PIOSP00008W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water.
This gene encodes a member of the polycystin protein family containing 11 transmembrane domains, a receptor for egg jelly (REJ) domain, and a polycystin-1, lipoxygenase, alpha-toxin (PLAT) domain. The encoded protein may play a role in the male reproductive system. Alternative splice variants have been described but their biological nature has not been determined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PKD1L1
Polyclonal
PKD1L1
PKD1L1, Polycystin-1L1, PK1L1
Rabbit
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q8TDX9
PKD1L1
A synthetic peptide from the 6th cytoplasmic loop of human Polycystic kidney disease protein 1-like 1
RUO
168507
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations
Recommendations