Learn More
PKD1L1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSP00013W
Catalog No. PIOSP00013W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
This gene encodes a member of the polycystin protein family containing 11 transmembrane domains, a receptor for egg jelly (REJ) domain, and a polycystin-1, lipoxygenase, alpha-toxin (PLAT) domain. The encoded protein may play a role in the male reproductive system. Alternative splice variants have been described but their biological nature has not been determined.
Specifications
Specifications
PKD1L1
Polyclonal
whole serum with no preservative
Q8TDX9
PKD1L1, Polycystin-1L1, PK1L1
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
PKD1L1
Lyophilized
PKD1L1
A synthetic peptide from the 1t extracellular loop of human Polycystic kidney disease protein 1-like 1
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
168507