Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-PKD2, Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ ST104250UG
Description
Specifically detects PKD2 Clone: in Human samples, and it is validated for Immunoprecipitation
Protein kinase D2 (PKD2) is a serine/threonine protein kinase and a member of PKD family, which also includes PKD/PKD1/PKCμ and PKD3/PKCν. PKDs are a newly identified protein kinase family with structural, enzymological and regulatory properties different from protein kinase C (PKC) family. PKDs participate in the signal transduction pathway and are potential regulators of cell proliferation and apoptosis. They are stimulated by diacyl-glycerol (DAG), and are substrates for classical PKCs like PKCα, PKCε and PKCη. Therefore, PKD stimulation by DAG is partially mediated through PKCs. PKD2 mRNA is widely expressed in human and murine tissue and codes for a∽105kDa protein. It is stimulated by both phorbol ester and growth factors and is autophosphorylated upon stimulation. PKD2 is predominantly located in the cytoplasm and, as opposed to other members of the PKD family, does not accumulate in the nucleus upon induction. This suggests that PKD2 mediates physiological responses different from other members of the PKD family.Specifications
PKD2 | |
Polyclonal | |
In PBS. | |
a synthetic peptide corresponding to amino acids near the C-terminus of human PKD2 | |
RUO | |
Recognizes the ∽105kDa PKD2 protein in HEK293 cells. | |
2°C to 8°C | |
IgG |
Immunoprecipitation | |
Unconjugated | |
Rabbit | |
50 μg | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction