Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cGKI Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128083

Catalog No. PA128083


Add to cart

Description

Description

PA1-28083 detects PKG from human, rat, bovine, and mouse samples. PA1-28083 has been successfully used in immunocytochemistry, immunohistochemistry, immunoprecipitation, and Western blot applications.A suggested positive control is HeLa cell lysate The PA1-28083 immunogen is a 14-residue synthetic peptide corresponding to residues 4-17 of human PKG 1 beta. Tryptophan was substituted for tyrosine at position 9. A terminal cysteine was added and the peptide conjugated to KLH. Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Specifications

Specifications

cGKI
Polyclonal
Unconjugated
PRKG1
Liquid
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
19091, 282004, 54286, 5592
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
P00516, Q13976, P0C605
PRKG1
A 14-residue synthetic peptide corresponding to residues 4-17 of human PKG 1 beta.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit