Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKHD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals BC11060479
Description
PKHD1 Polyclonal specifically detects PKHD1 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
PKHD1 | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
polycystic kidney and hepatic disease 1 (autosomal recessive) | |
Rabbit | |
Affinity Purified | |
RUO | |
5314 | |
Human, Mouse | |
IgG |
Immunocytochemistry, Immunofluorescence, ELISA | |
0.2 mg/mL | |
Immunocytochemistry/Immunofluorescence, Peptide ELISA 1:100-1:2000 | |
P08F94 | |
PKHD1 | |
A synthetic peptide derived from the human PKHD1 protein (within residues 3300-3400). [UniProt# Q8TCZ9] | |
0.1 mL | |
Primary | |
Human. Immunogen sequence has 83% identity to mouse. Customer review on mouse kidney collecting duct cell line. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction