Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKM2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 14 publications
Supplier: Novus Biologicals NBP148308SS
Description
PKM2 Polyclonal specifically detects PKM2 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Knockdown Validated.Specifications
PKM2 | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.1% Sodium Azide | |
CTHBP, Cytosolic thyroid hormone-binding protein, MGC3932, OIP-3, OIP3EC 2.7.1.40, Opa-interacting protein 3, PK, muscle type, PK2, PK3PKM, Pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, pyruvate kinase, muscle, TCB, THBP1p58, Thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, Tumor M2-PK | |
Rabbit | |
60 kDa | |
0.025 mL | |
Cancer, Lipid and Metabolism | |
5315 | |
Human, Mouse, Rat, Bovine | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
1.0 mg/ml | |
Western Blot 0.5 ug/ml, Simple Western 1:12.5, Flow Cytometry 5 ug/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation reported in scientific literature (PMID 21620138), Immunohistochemistry-Paraffin 1:100, Immunoblotting reported in scientific literature (PMID 25416385), KnockDown Validated | |
P14618, P52480 | |
PKM | |
A synthetic peptide made to the internal region of human PKM2 protein (within residues 350-450). [Swiss-Prot: P14618] | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse and rat. Immunogen sequence has 95% identity to bovine. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction