Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKM2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525962

Catalog No. PIPA525962

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse, rat and rabbit based on sequence homology.

PKM2 is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. PKM2 has been shown to interact with thyroid hormone, and thus may mediate cellular metabolic effects induced by thyroid hormones. PKM2 has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Three alternatively spliced transcript variants encoding two distinct isoforms of PKM2 have been reported.
Specifications

Specifications

PKM2
Polyclonal
PBS with 0.09% sodium azide
P14618
CTHBP, OIP3, PK3, PKM, TCB, THBP1, OIP-3, OPA-interacting protein 3, PK, muscle type, cytosolic thyroid hormone-binding protein, p58, pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, pyruvate kinase muscle isozyme, thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, tu
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5315
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
PKM
Liquid
PKM
KLH conjugated synthetic peptide between 116-145 amino acids from the N-terminal region of human PKM2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit