Learn More
PKR Mouse anti-Human, Unlabeled, Clone: 13, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610764
Description
Double stranded RNA (dsRNA) generated by most viruses during the infectious cycle is a potent stimulator of interferons. Interferons bind to cell surface receptors and stimulate the synthesis of several proteins that interfere with viral replication. One protein, 2′5′-oligo-adenylate synthetase, indirectly activates an endoribonuclease that degrades viral RNA. Another protein, p68 serine/threonine protein kinase (also known as PKR and TIK), is induced following interferon stimulation and activated by autophosphorylation in the presence of dsRNA. Upon activation, p68 phosphorylates the α subunit of the eukaryotic initiation factor 2 (eIF-2) resulting in inhibition of protein synthesis and, in turn, inhibition of viral replication. Evidence also suggests that p68 protein kinase inhibits proliferation and potentiates tumor suppressor function. In addition, p68 has been shown to phosphorylate Iκ-B, thus activating NF-κB which induces interferon-β gene transcription.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
PKR | |
Monoclonal | |
250μg/mL | |
Protein Kinase R; p68 Kinase; TIK | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
13 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human p68 Kinase aa. 117-250 | |
50 μg | |
Cell Biology | |
Human | |
IgG1 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.