Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA1A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526555

Catalog No. PIPA526555

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

Phosphatidylserine-specific phospholipase A1-alpha (PLA1A) acts specifically on phosphatidylserine (PS) and 1-acyl-2-lysophosphatidylserine (lyso-PS) to hydrolyze fatty acids at the sn-1 position of these phospholipids.
Specifications

Specifications

PLA1A
Polyclonal
PBS with 0.09% sodium azide
Q53H76, Q8VI78
PS-PLA1, PSPLA1, phosphatidylserine-specific phospholipase A1alpha
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51365, 85031
Western Blot
Unconjugated
PLA1A
Liquid
PLA1A
KLH conjugated synthetic peptide between 329-358 amino acids from the central region of human PLA1A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit