Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA2 Mouse anti-Human, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05143
Description
This Anti-PLA2 Antibody, secretory is validated for use in ELISA, immunohistochemistry, western blotting for the detection of PLA2.Specifications
PLA2 | |
Monoclonal | |
0.1M Tris-glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide | |
PLA2G2A, sPLA2, PLA2S, PLA2, MOM1, PLA2B, RASF-A, PLAS1, NPS-PLA2, PLA2L | |
Purified human PLA2; recognizes type II (secretory) PLA2. | |
200 μg | |
Human | |
Purified |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
NM_000300.2 | |
Mouse | |
DEAE cellulose chromatography | |
Primary | |
−20°C for 24 months. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only