Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527776

Catalog No. PIPA527776


Add to cart

Description

Description

PA5-27776 targets PLA2G1B in WB applications and shows reactivity with Human samples. The PA5-27776 immunogen is recombit fragment corresponding to a region within amino acids 1 and 148 of Human PLA2G1B. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Phospholipase A2 (EC 3.1.1.4) catalyzes the release of fatty acids from glycero-3-phosphocholines. The best known varieties are the digestive enzymes secreted as zymogens by the pancreas of mammals. Sequences of pancreatic PLA2 enzymes from a variety of mammals have been reported. One striking feature of these enzymes is their close homology to venom phospholipases of snakes. Other forms of PLA2 have been isolated from brain, liver, lung, spleen, intestine, macrophages, leukocytes, erythrocytes, inflammatory exudates, chondrocytes, and platelets (Seilhamer et al., 1986 [PubMed 3028739]) .
Specifications

Specifications

PLA2
Polyclonal
Unconjugated
PLA2G1B
Liquid
PLA2G1B
Recombit fragment corresponding to a region within amino acids 1 and 148 of Human PLA2G1B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.74 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P04054
PLA2, PLA2A, PPLA2, group IB phospholipase A2, phosphatidylcholine 2-acylhydrolase 1B, phospholipase A2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5319
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit