Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA524805
Description
PLA2G1B catalyzes the release of fatty acids from glycero-3-phosphocholines. The best known varieties are the digestive enzymes secreted as zymogens by the pancreas of mammals. Sequences of pancreatic PLA2 enzymes from a variety of mammals have been reported. One striking feature of these enzymes is their close homology to venom phospholipases of snakes. Other forms of PLA2 have been isolated from brain, liver, lung, spleen, intestine, macrophages, leukocytes, erythrocytes, inflammatory exudates, chondrocytes, and plateletsSpecifications
PLA2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P04054, Q9Z0Y2 | |
PLA2, PLA2A, PPLA2, group IB phospholipase A2, phosphatidylcholine 2-acylhydrolase 1B, phospholipase A2 | |
Rabbit | |
IgG | |
400 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
18778, 5319 |
Flow Cytometry, Western Blot | |
Unconjugated | |
PLA2G1B | |
Liquid | |
PLA2G1B | |
KLH conjugated synthetic peptide between 116-145 amino acids from the C-terminal region of human PLA2G1B | |
Antigen affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title