Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA2G4B Rabbit anti-Human, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA524844
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
PLA2G4B transcribes naturally-occurring mRNAs that are co-transcribed products of the neighboring JMJD7 and PLA2G4B genes. Incompletely processed read-through transcripts from these two loci are abundantly expressed in most tissues, but the function of the predicted protein product has not yet been determined.Specifications
PLA2G4B | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P0C869 | |
HsT16992, cPLA2-beta, cytosolic phospholipase A2 beta, phospholipase A2 group IVB | |
Rabbit | |
IgG | |
400 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
100137049 |
Flow Cytometry, Western Blot | |
Unconjugated | |
PLA2G4B | |
Liquid | |
PLA2G4B | |
KLH conjugated synthetic peptide between 396-424 amino acids from the central region of human PLA2G4B | |
Antigen affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title