Learn More
PLA2G6 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA526936
Catalog No. PIPA526936
Add to cart
Description
Description
The protein encoded by this gene is an A2 phospholipase, a class of enzyme that catalyzes the release of fatty acids from phospholipids. The encoded protein may play a role in phospholipid remodelling, arachidonic acid release, leukotriene and prostaglandin synthesis, fas-mediated apoptosis, and transmembrane ion flux in glucose-stimulated B-cells.
Specifications
Specifications
PLA2G6
Polyclonal
Unconjugated
PLA2G6
Liquid
PLA2G6
KLH conjugated synthetic peptide between 551-580 amino acids from the central region of human PLA2G6
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
2.0 mg/mL
PBS with 0.09% sodium azide
O60733
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, 85 kDa calcium-independent phospholipase A2, GVI PLA2, calcium-independent phospholipase A2, cytosolic, calcium-independent phospholipase A2, group VI phospholipase A2, patatin-like phospholipase domain containing 9
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8398