Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
pleiomorphic adenoma gene 1, Mouse, Clone: 3B7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PLAG1.
Supplier: Abnova Corporation H00005324M02
Description
Pleomorphic adenoma gene 1 encodes a zinc finger protein with 2 putative nuclear localization signals. PLAG1, which is developmentally regulated, has been shown to be consistently rearranged in pleomorphic adenomas of the salivary glands. PLAG1 is activated by the reciprocal chromosomal translocations involving 8q12 in a subset of salivary gland pleomorphic adenomas. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq
Sequence: ATVIPGDLSEVRDTQKVPSGKRKRGETKPRKNFPCQLCDKAFNSVEKLKVHSYSHTGERPYKCIQQDCTKAFVSKYKLQRHMATHSPEKTHKCNYCEKSpecifications
pleiomorphic adenoma gene 1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_002655 | |
PLAG1 | |
PLAG1 (NP_002646, 2 a.a. ∼ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Transcription Regulation | |
5324 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, Western Blot | |
3B7 | |
Mouse monoclonal antibody raised against a partial recombinant PLAG1. | |
PLAG1 | |
PSA/SGPA | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction