Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plakoglobin Mouse anti-Human, Clone: 15F11, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04139
Description
This Anti-Plakoglobin Antibody, clone 15F11 is validated for use in western blotting for the detection of Plakoglobin.Specifications
Plakoglobin | |
Monoclonal | |
Unconjugated | |
NP_002221 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Western Blot | |
15F11 | |
Purified mouse monoclonal IgG1κ in buffer containing PBS and 0.05% sodium azide. | |
JUP, CTNNG, DP3 | |
Recombinant protein of plakoglobin. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only