Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKP1 Mouse anti-Human, Clone: 10B2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA12004

Catalog No. 01-674-557

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.This product is either new or inactive, and cannot be ordered online.


Description

Description

Plakophilin-1 Antibody (10B2), MA1-2004, from Invitrogen. Species Reactivity: Human; Applications: Western Blot.

This gene encodes a member of the arm-repeat (armadillo) and plakophilin gene families. Plakophilin proteins contain numerous armadillo repeats, localize to cell desmosomes and nuclei, and participate in linking cadherins to intermediate filaments in the cytoskeleton. This protein may be involved in molecular recruitment and stabilization during desmosome formation. Mutations in this gene have been associated with the ectodermal dysplasia/skin fragility syndrome.
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG1
Clone: 10B2
Immunogen: The N-terminal region of human plakophilin-1.
Formulation: 200μg purified antibody (1mg/mL) in PBS with 1mg/mL BSA and 0.05% sodium azide
Synonyms: B6P, MGC138829
Uniprot Accession: Q13835
Entrez GeneID: 5317
Gene Symbol: PKP1
MA1-2004 contains 200 ug of in vitro produced, protein A purified antibody (1 mg/ml) in PBS containing 1 mg/ml BSA and 0.05% sodium azide. MA1-2004 detects plakophilin-1from human samples. MA1-2004 has been successfully used in Western blot applications. By Western blot, MA1-2004 detects a ∽70 kDa band representing plakophilin-1 from MCF-7 cells. The MA1-2004 immunogen is the N-terminal region of human plakophilin-1.
Specifications

Specifications

PKP1
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q13835
B6P, MGC138829
Mouse
IgG1
200 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5317
Western Blot
10B2
Unconjugated
PKP1
Liquid
PKP1
The N-terminal region of human plakophilin-1.
Protein A
RUO
Antibody
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit