Learn More
Urokinase Mouse anti-Human, Clone: U-16, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MONU1602
Catalog No. PIMONU1602
Add to cart
Description
Description
MON U-16-02 is specific for human uPA. No reaction with human tissue plasminogen activator (tPA) when tested by ELISA, immunoblotting and enzyme inhibition and no reaction with any other human plasma proteins when tested by immunoblotting. MON U-16-02 has successfully been used in ELISA, Western blot and immunohistochemistry (frozen) applications. The immunogen used to produce MON U-16-02 is native human uPA. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Urokinase plasminogen activator (uPA) is a serine protease. It converts the abundant proenzyme plasminogen to active plasmin and plays a key role in cancer invasion and a variety of tissue remodelling processes such as wound healing, mammary gland involution and placental development. Elevated levels of uPA are associated with poor prognosis in many types of cancer.
Specifications
Specifications
Urokinase
Monoclonal
1.10 mg/mL
PBS with 15mM sodium azide; pH 7.4
P00749
CD87, UPAR, URKR
Mouse
IgG1
200 μL
4° C, store in dark, DO NOT FREEZE!
Primary
5328
ELISA, Immunohistochemistry (Frozen), Western Blot
U-16
Unconjugated
PLAU
Liquid
PLAU
Native human uPA.
Protein A
RUO
Antibody
Monoclonal
Human